siRNA Details
| Gene Symbol: LINC01355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110616.1 | 1 | 234 | |||
| GenBank mRNA | AL832122.1 | - | 1 | 1468 | ||
| BX113324.1 | - | 1 | 234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01355| Gene Symbol: LINC01355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110616.1 | 1 | 234 | |||
| GenBank mRNA | AL832122.1 | - | 1 | 1468 | ||
| BX113324.1 | - | 1 | 234 | |||