siRNA Details
| Gene Symbol: LOC105378367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132791.1 | 2 | 358 | |||
| GenBank mRNA | HY007587.1 | - | 2 | 359 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000445111.1 | - | 1 | 103 | Chr. 10 - 75742740 - 75743755 [+] on Build GRCh38 | |

LOC105378367
Genomic Map