siRNA Details
| Gene Symbol: LINC01014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046786.1 | 3 | 398 | |||
| GenBank mRNA | BX105665.1 | - | 3 | 399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01014| Gene Symbol: LINC01014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046786.1 | 3 | 398 | |||
| GenBank mRNA | BX105665.1 | - | 3 | 399 | ||