siRNA Details
| Gene Symbol: ERICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017013135.1 | XP_016868624.1 | 1 | 573 | ||
| XM_017013136.1 | XP_016868625.1 | 1 | 573 | |||
| XM_017013137.1 | XP_016868626.1 | 1 | 573 | |||
| XM_017013138.1 | XP_016868627.1 | 1 | 573 | |||
| XM_017013139.1 | XP_016868628.1 | 1 | 573 | |||

ERICH1
Genomic Map