siRNA Details
| Gene Symbol: ERICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017013137.1 | XP_016868626.1 | 1, 2 | 622 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH1| Gene Symbol: ERICH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017013137.1 | XP_016868626.1 | 1, 2 | 622 | ||