siRNA Details
| Gene Symbol: LINC00437 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126377.1 | 3 | 849 | |||
| GenBank mRNA | BC040923.2 | - | 2 | 623 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM927018.1 | - | 655 | Chr. - - [-] on Build | ||

LINC00437
Genomic Map