siRNA Details
Gene Symbol: LOC101927526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109966.1 | 3 | 600 | |||
GenBank mRNA | BC035864.1 | - | 3 | 611 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI086904.1 | - | 3 | 600 | Chr. 22 - 44443327 - 44444790 [+] on Build GRCh38 |
