siRNA Details
| Gene Symbol: LOC339803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036496.1 | 2 | 2364 | |||
| GenBank mRNA | BC008254.1 | - | 1 | 859 | 3872056 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC339803| Gene Symbol: LOC339803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036496.1 | 2 | 2364 | |||
| GenBank mRNA | BC008254.1 | - | 1 | 859 | 3872056 | |