siRNA Details
| Gene Symbol: LOC339803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036496.1 | 2 | 2529 | |||
| GenBank mRNA | BC008254.1 | - | 1 | 1024 | 3872056 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420918.3 | - | 2 | 2504 | Chr. 2 - 61141597 - 61144950 [-] on Build GRCh38 | |

LOC339803
Genomic Map