siRNA Details
Gene Symbol: BSN-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038866.1 | 3 | 1979 | |||
GenBank mRNA | AK096387.1 | - | 3 | 1979 | ||
AK097709.1 | - | 4 | 1819 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BSN-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038866.1 | 3 | 1979 | |||
GenBank mRNA | AK096387.1 | - | 3 | 1979 | ||
AK097709.1 | - | 4 | 1819 |