siRNA Details
| Gene Symbol: HCG22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003948.2 | 3 | 938 | |||
| GenBank mRNA | AB600644.1 | - | 3 | 938 | ||
| AK094433.1 | - | 2 | 750 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCG22| Gene Symbol: HCG22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003948.2 | 3 | 938 | |||
| GenBank mRNA | AB600644.1 | - | 3 | 938 | ||
| AK094433.1 | - | 2 | 750 | |||