siRNA Details
| Gene Symbol: LOC105372626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134577.1 | 2 | 444 | |||
| GenBank mRNA | BG185028.1 | - | 1 | 377 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000428529.1 | - | 2 | 442 | Chr. 20 - 43894720 - 43895468 [+] on Build GRCh38 | |

LOC105372626
Genomic Map