siRNA Details
Gene Symbol: LINC01522 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110027.1 | 3 | 411 | |||
GenBank mRNA | DB066225.1 | - | 3 | 386 | ||
DB071041.1 | - | 3 | 379 | |||
DB088024.1 | - | 3 | 340 | |||
HY050016.1 | - | 3 | 412 |
