siRNA Details
| Gene Symbol: TUSC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104174.1 | 2 | 1006 | |||
| GenBank mRNA | BC025370.2 | - | 2 | 907 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432701.2 | - | 2 | 907 | Chr. 13 - 44400250 - 44405885 [-] on Build GRCh38 | |

TUSC8
Genomic Map