siRNA Details
| Gene Symbol: LINC00441 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046414.1 | 1 | 324 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000433480.2 | - | 1 | 324 | Chr. 13 - 48296513 - 48303661 [-] on Build GRCh38 | |

LINC00441
Genomic Map