siRNA Details
| Gene Symbol: LOC101928739 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125401.1 | 3 | 205 | |||
| GenBank mRNA | BC032034.1 | - | 3 | 205 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435560.1 | - | 3 | 205 | Chr. 3 - 179101366 - 179147973 [-] on Build GRCh38 | |

LOC101928739
Genomic Map