siRNA Details
| Gene Symbol: LINC00399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126361.1 | 1, 2 | 116 | |||
| GenBank mRNA | BX111569.1 | - | 1, 2 | 116 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439008.1 | - | 1, 2 | 116 | Chr. 13 - 109400696 - 109401641 [+] on Build GRCh38 | |

LINC00399
Genomic Map