siRNA Details
| Gene Symbol: FOCAD-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121601.1 | 2 | 234 | |||
| GenBank mRNA | BX115825.1 | - | 2 | 234 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | F21658.1 | - | 2, 3 | 213 | Chr. 12 - 52233244 - 52248917 [+] on Build GRCh38 | |

FOCAD-AS1
Genomic Map