siRNA Details
Gene Symbol: LOC102723649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125867.1 | 3, 4 | 349 | |||
GenBank mRNA | BX098901.1 | - | 3, 4 | 364 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000440397.1 | - | 2, 3 | 352 | Chr. 6 - 137673460 - 137674554 [+] on Build GRCh38 |
