siRNA Details
| Gene Symbol: LOC102723649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125867.1 | 3 | 202 | |||
| GenBank mRNA | BX098901.1 | - | 3 | 217 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723649| Gene Symbol: LOC102723649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125867.1 | 3 | 202 | |||
| GenBank mRNA | BX098901.1 | - | 3 | 217 | ||