siRNA Details
| Gene Symbol: LOC102723649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125867.1 | 3 | 266 | |||
| GenBank mRNA | BX098901.1 | - | 3 | 281 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000440397.1 | - | 2 | 269 | Chr. 6 - 137673460 - 137674554 [+] on Build GRCh38 | |

LOC102723649
Genomic Map