siRNA Details
| Gene Symbol: LOC105376351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136146.1 | 3 | 264 | |||
| GenBank mRNA | BX095760.1 | - | 3 | 238 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441907.1 | - | 2 | 177 | Chr. 10 - 2446253 - 2447352 [-] on Build GRCh38 | |

LOC105376351
Genomic Map