siRNA Details
Gene Symbol: LINC00380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104057.1 | 2 | 66 | |||
GenBank mRNA | BX097335.1 | - | 2 | 66 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000442478.1 | - | 1 | 21 | Chr. 13 - 91087254 - 91089593 [-] on Build GRCh38 |
