siRNA Details
| Gene Symbol: LINC00529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ301561.1 | - | 2 | 149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00529| Gene Symbol: LINC00529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ301561.1 | - | 2 | 149 | ||