siRNA Details
Gene Symbol: LOC105372672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134576.1 | 1 | 138 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000444112.1 | - | 1 | 138 | Chr. 20 - 53608354 - 53634590 [+] on Build GRCh38 |
