siRNA Details
| Gene Symbol: LOC101929697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134939.1 | 2 | 163 | |||
| NR_134940.1 | 2 | 127 | ||||
| GenBank mRNA | BI042007.1 | - | 2 | 182 | ||
| DA288418.1 | - | 2 | 130 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG197322.1 | - | 2 | 108 | Chr. 3 - 195836470 - 195854209 [-] on Build GRCh38 | |

LOC101929697
Genomic Map