siRNA Details
Gene Symbol: LINC01011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055096.1 | - | 1 | 714 | ||
AK123663.1 | - | 2 | 1050 | |||
BC137372.1 | - | 1 | 616 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055096.1 | - | 1 | 714 | ||
AK123663.1 | - | 2 | 1050 | |||
BC137372.1 | - | 1 | 616 |