siRNA Details
| Gene Symbol: LOC101927532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135116.1 | 2 | 642 | |||
| GenBank mRNA | BG181918.1 | - | 1 | 365 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927532| Gene Symbol: LOC101927532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135116.1 | 2 | 642 | |||
| GenBank mRNA | BG181918.1 | - | 1 | 365 | ||