siRNA Details
| Gene Symbol: LOC105376955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136189.1 | 1 | 40 | |||
| GenBank mRNA | BG759368.1 | - | 1 | 41 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376955| Gene Symbol: LOC105376955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136189.1 | 1 | 40 | |||
| GenBank mRNA | BG759368.1 | - | 1 | 41 | ||