siRNA Details
| Gene Symbol: LOC105373185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135616.1 | 2 | 231 | |||
| GenBank mRNA | BE613553.1 | - | 2 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105373185| Gene Symbol: LOC105373185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135616.1 | 2 | 231 | |||
| GenBank mRNA | BE613553.1 | - | 2 | 231 | ||