siRNA Details
| Gene Symbol: LOC105377213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135614.1 | 2 | 438 | |||
| GenBank mRNA | BX103087.1 | - | 2 | 438 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000454307.1 | - | 2 | 438 | Chr. X - 151515551 - 151516245 [+] on Build GRCh38 | |

LOC105377213
Genomic Map