siRNA Details
Gene Symbol: LINC00885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034088.1 | 3 | 1313 | |||
GenBank mRNA | AK129507.1 | - | 3 | 1313 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000457079.1 | - | 3 | 1313 | Chr. 3 - 196142636 - 196160890 [+] on Build GRCh38 |
