siRNA Details
Gene Symbol: LINC00885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034088.1 | 1 | 192 | |||
GenBank mRNA | AK129507.1 | - | 1 | 192 | ||
BC144578.1 | - | 1 | 73 | |||
BC144579.1 | - | 1 | 73 | |||
BC144580.1 | - | 1 | 73 | |||
BC151150.1 | - | 1 | 73 | 9021323 |
