siRNA Details
| Gene Symbol: LINC01436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110419.1 | 2 | 571 | |||
| GenBank mRNA | CF127993.1 | - | 2 | 553 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU595315.1 | - | 40 | Chr. - - [-] on Build | ||

LINC01436
Genomic Map