siRNA Details
| Gene Symbol: LOC101927740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109890.1 | 3 | 2449 | |||
| GenBank mRNA | AK123543.1 | - | 3 | 2249 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927740| Gene Symbol: LOC101927740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109890.1 | 3 | 2449 | |||
| GenBank mRNA | AK123543.1 | - | 3 | 2249 | ||