siRNA Details
| Gene Symbol: LOC728339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC087857.1 | - | 3 | 634 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000501825.2 | - | 3 | 631 | Chr. 4 - 189864262 - 189939837 [-] on Build GRCh38 | |

LOC728339
Genomic Map