siRNA Details
Gene Symbol: LINC01596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132380.1 | 1 | 190 | |||
GenBank mRNA | BX094246.1 | - | 1 | 190 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000503609.1 | - | 1 | 190 | Chr. 4 - 189881515 - 189884868 [+] on Build GRCh38 |
