siRNA Details
Gene Symbol: LOC101929019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125910.1 | 4 | 548 | |||
GenBank mRNA | BC042433.1 | - | 4 | 519 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG723674.1 | - | 4 | 523 | Chr. 4 - 12223482 - 12251260 [+] on Build GRCh38 |
