siRNA Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006714446.3 | XP_006714509.1 | 3 | 588 | ||
| XM_011513968.2 | XP_011512270.1 | 3 | 588 | |||
| XM_011513969.2 | XP_011512271.1 | 3 | 465 | |||
| XM_011513970.1 | XP_011512272.1 | 3 | 177 | |||
| XM_011513971.2 | XP_011512273.1 | 3 | 533 | |||
| XM_011513972.2 | XP_011512274.1 | 3 | 589 | |||
| XM_011513973.1 | XP_011512275.1 | 3 | 586 | |||
| XM_017009112.1 | XP_016864601.1 | 3 | 589 | |||

PLEKHG4B
Genomic Map