siRNA Details
| Gene Symbol: LOC102577426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108031.1 | 2 | 337 | |||
| GenBank mRNA | HY011707.1 | - | 2 | 338 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000513771.1 | - | 2 | 281 | Chr. 5 - 181191046 - 181191852 [-] on Build GRCh38 | |

LOC102577426
Genomic Map