siRNA Details
| Gene Symbol: LOC102577426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108031.1 | 2 | 411 | |||
| GenBank mRNA | HY011707.1 | - | 2 | 412 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102577426| Gene Symbol: LOC102577426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108031.1 | 2 | 411 | |||
| GenBank mRNA | HY011707.1 | - | 2 | 412 | ||