siRNA Details
| Gene Symbol: MIR124-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034103.2 | 1 | 127 | |||
| NR_109793.1 | 1 | 121 | ||||
| GenBank mRNA | BM717762.1 | - | 1 | 127 | ||
| BQ639660.1 | - | 1 | 121 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR124-2HG| Gene Symbol: MIR124-2HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034103.2 | 1 | 127 | |||
| NR_109793.1 | 1 | 121 | ||||
| GenBank mRNA | BM717762.1 | - | 1 | 127 | ||
| BQ639660.1 | - | 1 | 121 | |||