siRNA Details
Gene Symbol: LOC105369423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135090.1 | 1 | 334 | |||
GenBank mRNA | BX098897.1 | - | 1 | 339 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105369423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135090.1 | 1 | 334 | |||
GenBank mRNA | BX098897.1 | - | 1 | 339 |