siRNA Details
Gene Symbol: LINC01479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM850770.1 | - | 3 | 421 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM854577.1 | - | 3 | 421 | Chr. 12 - 67929235 - 67932942 [+] on Build GRCh38 |
