siRNA Details
| Gene Symbol: LINC01315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120595.1 | 2 | 1122 | |||
| NR_120596.1 | 2 | 703 | ||||
| NR_120597.1 | 2 | 360 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01315| Gene Symbol: LINC01315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120595.1 | 2 | 1122 | |||
| NR_120596.1 | 2 | 703 | ||||
| NR_120597.1 | 2 | 360 | ||||