siRNA Details
| Gene Symbol: LOC100507406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121618.1 | 3 | 711 | |||
| NR_121619.1 | 2 | 677 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507406| Gene Symbol: LOC100507406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121618.1 | 3 | 711 | |||
| NR_121619.1 | 2 | 677 | ||||