siRNA Details
| Gene Symbol: LOC100507406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121618.1 | 1 | 190 | |||
| NR_121619.1 | 1 | 190 | ||||
| GenBank mRNA | BF970945.1 | - | 1 | 191 | ||
| N41668.1 | - | 1 | 136 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507406| Gene Symbol: LOC100507406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121618.1 | 1 | 190 | |||
| NR_121619.1 | 1 | 190 | ||||
| GenBank mRNA | BF970945.1 | - | 1 | 191 | ||
| N41668.1 | - | 1 | 136 | |||