siRNA Details
| Gene Symbol: RALY-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109885.1 | 3 | 379 | |||
| NR_109886.1 | 2 | 288 | ||||
| GenBank mRNA | AA447916.1 | - | 3 | 362 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419662.2 | - | 2 | 278 | Chr. 20 - 33992511 - 33994357 [-] on Build GRCh38 | |

RALY-AS1
Genomic Map