siRNA Details
| Gene Symbol: LOC100506470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109919.1 | 1 | 186 | |||
| GenBank mRNA | BX106486.1 | - | 1 | 131 | ||
| HY047124.1 | - | 1 | 186 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441660.1 | - | 1 | 110 | Chr. 20 - 61079140 - 61080179 [+] on Build GRCh38 | |

LOC100506470
Genomic Map