siRNA Details
Gene Symbol: LOC100506271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110515.1 | 1, 2 | 1718 | |||
GenBank mRNA | AK097787.1 | - | 1, 2 | 1520 | ||
BC043218.2 | - | 1, 2 | 1718 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000445088.1 | - | 1, 2 | 1718 | Chr. 22 - 37352190 - 37354839 [-] on Build GRCh38 |
